Learn More
IL-21R Mouse anti-Human, PE, Clone: 17A12, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 560264
Description
IL-21 receptor (IL-21R) encodes a 538 amino acid cytokine receptor with an extracellular domain consisting of one copy of the conserved WSXWS -containing cytokine-binding domain. The IL-21 receptor combines with the common cytokine-receptor γ-chain to form a functional receptor for IL-21. IL-21 is mainly produced by CD4+ T cells. IL-21R is preferentially expressed by B cells, T cells, NK cells, some populations of myeloid cells, keratinocytes, and dendritic cells. Binding of its ligand, IL-21, in these cells results in the activation of the Jak/Stat signal transduction pathway. The effects IL-21 ligand binding has pleiotropic actions such as augmenting the proliferation of T cells, driving of B cells into memory cells, terminally differentiating plasma cells and augmenting the activity of natural killer cells. IL-21 receptor has anti-tumor activity and might have a role in the development of autoimmunity; it has been reported that the IL-21 receptor affects the homeostasis of regulatory T cells and it could enhance T cell-activated responses in human immune-inflammatory diseases.
Host Species: Mouse
Clone: 17A12
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry

Specifications
IL-21R | |
Monoclonal | |
PE | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
17A12 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human IL-21R Transfected Cell Line | |
100 Tests | |
Immunology | |
Human | |
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.