Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-23R Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. 061331MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-133-1MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-133-1MI Supplier MilliporeSigma Supplier No. 061331
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-IL-23R Antibody, cytoplasmic detects level of IL-23R and has been published and validated for use in western blot, immunohistochemistry, ELISA, flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-23R
Applications ELISA, Flow Cytometry, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in PBS with 0.05% sodium azide.
Gene Symbols il23r; IBD17; IL-23R
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human IL-23R at and around the cytoplasmic domain.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_653302
Target Species Human, Mouse
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.