Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-27 p28 (IL-30) Mouse anti-Mouse, NA/LE, Unlabeled, Clone: MM27.7B1, BD

Catalog No. BDB563284 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563284 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563284 Supplier BD Biosciences Supplier No. 563284
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MM27.7B1 monoclonal antibody specifically binds to the p28 subunit of mouse Interleukin- 27 (IL-27 p28) that is also known as IL-27 alpha (IL-27α) or Interleukin-30 (IL-30). IL-27 p28 is a member of the IL-6/IL-12 family of cytokines. The IL-27 p28 subu nit binds to the EBI3 (Epstein-Barr virus induced gene 3) protein to form heterodimeric Interleukin-27. IL-27 p28 is produced by macrophages and dendritic cells stimulated by inflammatory cytokines and ligands for Toll-like receptors (TLR). IL-27 binds to and transduces signal through surface IL-27 receptors (IL-27R) that are expressed by B cells and naοve T cells and more highly expressed by NK cells and activated T cells. The IL-27R complex is comprised of gp130 (CD130) and WSX-1 (IL-27R alpha) subunits. IL-27 signals through IL-27R that activate the Jak/STAT signaling pathway resulting in the phosphorylation of Stat1 and Stat3. It acts in a proinflammatory manner to upregulate cellular T-bet transcription fac tor and surface IL-12R beta2 expression contributing to higher levels of IFN-γand granzyme B expression. IL-27 exerts anti-inflammatory responses by suppressing the differentiation and proliferation of Th2 and Th17 cells. IL-27 p28 is reportedly produced and secreted independently of EBI3. It may inhibit the actions of IL-17 and other proinflammatory cytokines like IL-6. The MM27/7B1 antibody has been reported to neutralize IL-27 activity in vitro. Routine quality testing of mAb MM27.7B1 is performed via flow cytometry of intracellularly stained IL-27 p28-transfected cells.

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-27 p28 (IL-30)
Applications Flow Cytometry
Classification Monoclonal
Clone MM27.7B1
Concentration 1.0mg/mL
Conjugate Unconjugated
Description Il27; IL-27 alpha; Il27a; IL27-A; IL-27-A; IL-27α; IL-27 p28; Il30; IL-30
Formulation No azide/low endotoxin: Aqueous buffered solution containing no preservative, 0.2μm sterile filtered. Endotoxin level is ≤0.01 EU/μg (≤0.001 ng/μg) of protein as determined by the LAL assay.
Host Species Mouse
Immunogen Mouse IL-27
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.