Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-27Rα Rat anti-Mouse, PE, Clone: 2918, BD

Catalog No. BDB564337 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564337 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564337 Supplier BD Biosciences Supplier No. 564337
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 2918 monoclonal antibody specifically binds to the alpha subunit (IL-27 Receptor alpha/IL-27 R α ) of the mouse Interleukin-27 Receptor complex. IL-27 R α is a type I transmembrane glycoprotein that belongs to the type I cytokine receptor family. It is also known as WSX-1 and T cell cytokine receptor (TCCR). IL-27 R α combines with gp130 (CD130) to form a high-affinity cell surface receptor that transduces intracellular signals through the JAK-STAT pathway upon IL-27 binding. IL-27 is a heterodimeric cytokine that is composed of the Epstein-Barr virus-induced gene 3 (EBI3) and IL-27p28 (IL-30) protein subunits. This multifunctional cytokine is produced by activated macrophages, dendritic cells, and epithelial cells. It mediates a broad array of pro- and anti-inflammatory responses. The IL-27 Receptor is expressed by a variety of cell types including naοve T cells and B cells. It is more highly expressed by NK cells and subsets of activated T cells.

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-27Rα
Applications Flow Cytometry
Classification Monoclonal
Clone 2918
Concentration 0.2mg/mL
Conjugate PE
Description Il27ra; IL-27-R; IL-27Rα; T cell cytokine receptor; TCCR; WSX-1; CRL1
Host Species Rat
Immunogen Mouse TSRR Transfected Cell Line
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.