Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-IL-3 R beta, PE, Clone: 130705, R&D Systems™

Catalog No. FAB5492P
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB5492P 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB5492P Supplier R&D Systems Supplier No. FAB5492P
This item is not returnable. View return policy

Applications:

Flow Cytometry

Specifications

Antigen IL-3 R beta
Applications Flow Cytometry
Classification Monoclonal
Clone 130705
Conjugate R-PE
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. P26954
Gene Alias AIC2A, BetaIl3, Bil3, Csfgmrb, IL3R, IL3R beta, IL-3Rb, IL3RB, Il3rb2
Gene Symbols Csf2rb2
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse IL-3 R beta , His23-Trp440, Accession # P26954
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1439
Test Specificity Detects mouse IL-3 R beta in direct ELISAs. Cross-reactivity with AIC2B was not tested.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.