Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IL-34 PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Supplier: R&D Systems IC5265P
This item is not returnable.
View return policy
Description
IL-34 Monoclonal specifically detects IL-34 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
| IL-34 | |
| Monoclonal | |
| R-PE | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| C16orf77, IL34, IL-34, interleukin 34 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 146433 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 578416 | |
| Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
| Q6ZMJ4 | |
| IL34 | |
| Mouse myeloma cell line NS0-derived recombinant human IL-34 Asn21-Pro242 Accession # Q6ZMJ4 | |
| 100 Tests | |
| Primary | |
| Detects recombinant human IL-34 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse IL-34 is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction