Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-IL-37/IL-1F7, Alexa Fluor 488, Clone: 899826, R&D Systems™
Supplier: R&D Systems IC19751G025
This item is not returnable.
View return policy
Specifications
IL-37/IL-1F7 | |
Monoclonal | |
Alexa Fluor 488 | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
FIL1, FIL1 zeta, FIL1(ZETA), FIL1ZFIL1 zeta, IL-1F7, IL-1F7IL-1 zeta, IL1H4IL1F7 (canonical product IL-1F7b), IL-1H4IL-1F7b (IL-1H4, IL-1H, IL-1RP1), IL-1RP1IL-1X protein, IL1RP1interleukin 1 family member 7, IL-1X, IL37, interleukin 1 family, member 7 (zeta), interleukin 1, zeta, interleukin-1 family member 7, Interleukin-1 homolog 4, interleukin-1 superfamily z, Interleukin-1 zeta, Interleukin-1-related protein | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
27178 | |
Human | |
Purified |
Flow Cytometry | |
899826 | |
Intracellular Staining by Flow Cytometry 5 uL/10^6 cells | |
Q9NZH6 | |
IL37 | |
E. coli-derived recombinant human IL-37/IL-1F7, Lys53-Val206, Accession # Q9NZH6 | |
25 Tests | |
Primary | |
Detects human IL-37/IL-1F7 in direct ELISAs. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction