Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-4 Rat anti-Mouse, Alexa Fluor 488, Clone: 11B11, BD

Catalog No. BDB557728 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB557728 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB557728 Supplier BD Biosciences Supplier No. 557728
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 11B11 antibody reacts with mouse interleukin-4 (IL-4). The immunogen used to generate the 11B11 hybridoma was partially purified mouse IL-4 from PMA-stimulated EL-4 supernatant. This is a neutralizing antibody. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Rat
Clone: 11B11
Isotype: IgG1
Cross Reactivity: Mouse

Blocking Assay, Intracellular Staining

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-4
Applications Flow Cytometry
Classification Monoclonal
Clone 11B11
Concentration 0.2mg/mL
Conjugate Alexa Fluor 488
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Partially Purified Mouse IL-4
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.