Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-IL-4, Fluorescein, Clone: 3007, R&D Systems™

Catalog No. IC204F025
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC204F025 25 Tests
IC204F100 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. IC204F025 Supplier R&D Systems Supplier No. IC204F025
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 3 publications

IL-4 Monoclonal specifically detects IL-4 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen IL-4
Applications Flow Cytometry
Classification Monoclonal
Clone 3007
Conjugate Fluorescein
Dilution Intracellular Staining by Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. P05112.1
Gene Alias B cell growth factor 1, BCDF, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL4, IL-4B_cell stimulatory factor 1, IL4E12, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra
Gene Symbols IL4
Host Species Mouse
Immunogen E. coli-derived recombinant human IL-4 His25-Ser153 Accession # P05112.1
Purification Method Protein A or G purified from ascites
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3565
Test Specificity Detects human IL-4 in direct ELISAs and Western blots. In Western blots, this antibody does not cross-react with recombinant mouse IL-4.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.