Learn More
IL-4 Rat anti-Human, PE-CF594, Clone: MP4-25D2, BD
Rat Monoclonal Antibody
Supplier: BD Biosciences 565161
Description
The MP4-25D2 monoclonal antibody specifically binds to Interleukin-4 (IL-4). IL-4 is also known as Lymphocyte stimulatory factor 1, B cell stimulatory factor 1 (BSF-1), or B cell growth factor 1 (BCGF-1). IL-4 is produced by activated T cells, basophils, and mast cells. IL-4 is a multifunctional cytokine and growth factor that affects a variety of different target cell types. IL-4 can costimulate T cell proliferation and survival, as well as help drive Th2-like cell differentiation and effector functions. It costimulates B cell proliferation and survival, and can help B cells differentiate into IgG4- or IgE-producing cells. IL-4 can also diminish the proinflammatory functions of monocytes and macrophages. IL-4 exerts its biological effects by binding with high affinity to cell surface CD124 (IL-4R α chain). CD124 forms signaling IL-4 receptor complexes with either CD132/common γchain or CD213a1/IL-13R α 1 to form either Type I or Type II IL-4 Receptor complexes, respectively. The immunogen used to generate the MP4-25D2 hybridoma was purified recombinant human IL-4. This is a neutralizing antibody. The MP4-25D2 antibody has been reported to crossreact with IL-4 from rhesus monkeys. The use of the MP4-25D2 antibody for epitope mapping of human IL-4 has been described.

Specifications
IL-4 | |
Monoclonal | |
PE-CF594 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide | |
Purified Recombinant Human IL-4 | |
100 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
MP4-25D2 | |
Interleukin-4; IL4; BCGF-1; BSF-1; Lymphocyte stimulatory factor 1 | |
Rat | |
Affinity Purified | |
RUO | |
Human | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.