Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-4 Rat anti-Human, PE, Clone: MP4-25D2, BD

Catalog No. BDB554485 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554485 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554485 Supplier BD Biosciences Supplier No. 554485
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The MP4-25D2 antibody reacts with human interleukin-4 (IL-4). The immunogen used to generate the MP4-25D2 hybridoma was purified recombinant human IL-4. This is a neutralizing antibody. The MP4-25D2 antibody has been reported to cross react with IL-4 from rhesus monkeys. The use of the MP4-25D2 antibody for epitope mapping of human IL-4 has been described.

Host Species: Rat
Clone: MP4-25D2
Isotype: IgG1
Species Reactivity: Human
Immunogen: Purified Recombinant Human IL-4

Intracellular Staining

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-4
Applications Flow Cytometry
Classification Monoclonal
Clone MP4-25D2
Concentration 0.2mg/mL
Conjugate PE
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Purified Recombinant Human IL-4
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.