Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-5 Rat anti-Human, NA/LE, Unlabeled, Clone: JES1-5A10, BD

Catalog No. BDB562175 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562175 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562175 Supplier BD Biosciences Supplier No. 562175
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The JES1-5A10 monoclonal antibody specifically binds to the human Interleukin-5 (IL-5). IL-5 is a cytokine that is also known as T-cell replacing factor (TRF), Eosinophil differentiation factor (EDF), and B cell differentiation factor I. IL-5 is produced by T cells, mast cells, and eosinophils. IL-5 controls the generation, differentiation, activation and localization of eosinophils. IL-5 also activates and promotes the differentiation of basophils. IL-5 plays an important role in regulating cell-mediated immunity against parasitic infections and some tumors. High level IL-5 production is involved in some allergic diseases including asthma. IL-5 exerts its biological effects by binding to and signaling through high-affinity heterodimeric IL-5 receptors comprised of transmembrane IL-5 receptor alpha (IL-5R α, aka, CD125) and beta common (βc, aka, CD131) chains. The βc chain serves as the common signaling subunit for the IL-5 receptor heterodimer as well as for the heterodimeric IL-3 and GM-CSF receptors. The immunogen used to generate the JES1-5A10 hybridoma was recombinant human IL-5. The JES1-5A10 antibody has been reported to cross react with IL-5 from rhesus monkey. This is a neutralizing antibody.

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-5
Applications ELISA
Classification Monoclonal
Clone JES1-5A10
Concentration 1.0mg/mL
Conjugate Unconjugated
Description EDF; Eosinophil differentiation factor; TRF; T-cell replacing factor
Formulation No azide/low endotoxin: Aqueous buffered solution containing no preservative, 0.2μm sterile filtered. Endotoxin level is ≤0.01 EU/μg (≤0.001 ng/μg) of protein as determined by the LAL assay.
Host Species Rat
Immunogen Recombinant Human IL-5
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.