Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-6 Rat anti-Human, APC, Clone: MQ2-13A5, BD

Catalog No. BDB561441 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561441 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB561441 Supplier BD Biosciences Supplier No. 561441
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The MQ2-13A5 monoclonal antibody specifically binds to human interleukin-6 (IL-6). IL-6 is a multifunctional cytokine that plays a central role in host defense mechanisms, including hematopoiesis, immune responses (eg, T and B cell activation and differentiation) and acute phase reactions. IL-6 can be expressed by a variety of cells including monocytes/macrophages, eosinophils, fibroblasts, vascular endothelial cells, bone marrow stromal cells, mesangial cells, hepatocytes, keratinocytes, astrocytes, T lymphocytes, B lymphocytes, and various tumor cells. IL-6 production is upregulated by cells in response to bacterial products such as lipopolysaccharide, viruses and other pro-inflammatory cytokines such as IL-1, TNF , and IFN-γ. IL-6 transcription is downregulated by cells in response to IL-4 and IL-10. The functional IL-6 Receptor (IL-6R) complex consists of two transmembrane glycoproteins, an 80-kDa low-affinity ligand-binding receptor subunit (IL-6Rα/CD126) and a 130 kDa (gp130/CD130) subunit that binds to IL-6-IL-6Rα to form the high-affinity signal transducing complex. Abnormal expression of IL-6 is related to the pathogenesis of many diseases including neoplastic (eg, multiple myeloma) and autoimmune diseases (eg, rheumatoid arthritis). The immunogen used to generate this hybridoma was COS-7 -expressed recombinant human IL-6.

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-6
Applications Flow Cytometry
Classification Monoclonal
Clone MQ2-13A5
Conjugate APC
Description IL6; Interleukin-6; BSF-2; CDF; HGF; HSF; IFNB2
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Rat
Immunogen Human IL-6 Recombinant Protein
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.