Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-6 Rat anti-Mouse, NA/LE, Unlabeled, Clone: MP5-20F3, BD
Rat Monoclonal Antibody
Supplier: BD Biosciences 554398
Description
The MP5-20F3 antibody reacts with mouse interleukin-6 (IL-6). The immunogen used to generate the MP5-20F3 hybridoma was recombinant mouse IL-6. This is a neutralizing antibody.

Specifications
IL-6 | |
Monoclonal | |
1.0mg/mL | |
Il6; Il-6; Interleukin-6; B-cell hybridoma growth factor | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
ELISA | |
MP5-20F3 | |
Unconjugated | |
No azide/low endotoxin: Aqueous buffered solution containing no preservative, 0.2μm sterile filtered. Endotoxin level is ≤0.01 EU/μg (≤0.001 ng/μg) of protein as determined by the LAL assay. | |
Mouse IL-6 Recombinant Protein | |
0.5 mg | |
Immunology | |
Mouse | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction