Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-6 Rat anti-Human, Unlabeled, Clone: MQ2-6A3, BD
Rat Monoclonal Antibody
Supplier: BD Biosciences 559068
Description
The MQ2-6A3 antibody reacts with human interleukin-6 (IL-6). The immunogen used to generate the MQ2-6A3 hybridoma was recombinant human IL-6. This is a neutralizing antibody.
Immunocytochemistry

Specifications
IL-6 | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Recombinant human IL-6 | |
0.25 mg | |
Immunology | |
Human | |
IgG2a κ |
Immunocytochemistry | |
MQ2-6A3 | |
Unconjugated | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction