Learn More
IL-6 Rat anti-Human, PE-CF594, Clone: MQ2-13A5, BD
Rat Monoclonal Antibody
Supplier: BD Biosciences 563543
Description
The MQ2-13A5 monoclonal antibody specifically binds to human interleukin-6 (IL-6). IL-6 is a multifunctional cytokine that plays a central role in host defense mechanisms, including hematopoiesis, immune responses (eg, T and B cell activation and differentiation) and acute phase reactions. IL-6 can be expressed by a variety of cells including monocytes/macrophages, eosinophils, fibroblasts, vascular endothelial cells, bone marrow stromal cells, mesangial cells, hepatocytes, keratinocytes, astrocytes, T lymphocytes, B lymphocytes, and various tumor cells. IL-6 production is upregulated by cells in response to bacterial products such as lipopolysaccharide, viruses and other pro-inflammatory cytokines such as IL-1, TNF , and IFN-γ. IL-6 transcription is downregulated by cells in response to IL-4 and IL-10. The functional IL-6 Receptor (IL-6R) complex consists of two transmembrane glycoproteins, an 80-kDa low-affinity ligand-binding receptor subunit (IL-6Rα/CD126) and a 130 kDa (gp130/CD130) subunit that binds to IL-6-IL-6Rα to form the high-affinity signal transducing complex. Abnormal expression of IL-6 is related to the pathogenesis of many diseases including neoplastic (eg, multiple myeloma) and autoimmune diseases (eg, rheumatoid arthritis). The immunogen used to generate this hybridoma was COS-7 -expressed recombinant human IL-6.
Specifications
| IL-6 | |
| Monoclonal | |
| PE-CF594 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Human IL-6 Recombinant Protein | |
| 50 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| MQ2-13A5 | |
| IL6; Interleukin-6; BSF-2; CDF; HGF; HSF; IFNB2 | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.