Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-6 Mouse anti-Human, PE, Clone: AS12, BD

Catalog No. BDB340527 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB340527 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB340527 Supplier BD Biosciences Supplier No. 340527
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The Anti-Hu–IL-6 antibody, clone AS12, is derived from the fusion of P3X63.Ag8.653 myeloma cells with splenocytes from BALB/c mice immunized with recombinant human IL-6.

The Anti-Human Interleukin-6 (Anti-Hu–IL-6) antibody recognizes a 21- to 29-kilodalton (kDa) polypeptide.

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-6
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone AS12
Concentration 1.5μg/mL
Conjugate PE
Formulation Phosphate buffered saline with gelatin and 0.1% sodium azide.
Host Species Mouse
Immunogen Recombinant human IL-6 receptor antagonist
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.