Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-6 Rat anti-Human, PE, Clone: MQ2-6A3, BD

Catalog No. BDB551473 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB551473 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB551473 Supplier BD Biosciences Supplier No. 551473
Add to Cart
Add to Cart

Rat Monoclonal Antibody

IL-6, a 26kDa protein, is a multifunctional cytokine secreted by both lymphoid and non-lymphoid cells. It regulates B and T cell function and hematopoiesis. The MQ2-6A3 antibody reacts with human interleukin-6 (IL-6). The immunogen used to generate the MQ2-6A3 hybridoma was recombinant human IL-6. This is a neutralizing antibody. Clone MQ2-6A3 cross-reacts with an intracellular component of peripheral blood LPS-stimulated permeabilized monocytes of rhesus and cynomolgus macaques. The reactivity pattern observed on non-human primates is similar, although weaker, than that seen on normal human monocytes.

Host Species: Rat
Clone: MQ2-6A3
Isotype: IgG2a κ
Species Reactivity: Non-human primate
Immunogen: Recombinant human IL-6

Flow Cytometry, Intracellular Staining

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-6
Applications Flow Cytometry
Classification Monoclonal
Clone MQ2-6A3
Conjugate PE
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Rat
Immunogen Recombinant human IL-6
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Cynomolgus, Rhesus Monkey
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.