Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-IL-6, Recombinant rat, Biotin, Clone: G307-2, BD
Supplier: BD Biosciences 557008
Description
The G307-2 antibody reacts with rat IL-6. The immunogen used to generate the G307-2 hybridoma was recombinant rat IL-6.
Host Species: Mouse
Clone: G307-2
Isotype: IgG1, κ
Species Reactivity: Rat
Immunogen: Recombinant Rat IL-6
ELISA Detection, Western Blotting

Specifications
Primary | |
Rat |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction