Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-7 Rat anti-Human, Biotin, Clone: BVD10-11C10, BD

Catalog No. BDB554494 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554494 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554494 Supplier BD Biosciences Supplier No. 554494
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The BVD10-11C10 antibody reacts with human interleukin-7 (IL-7). The immunogen used to generate the BVD10- 11C10 hybridoma was recombinant human IL-7. This is a non-neutralizing antibody.

Host Species: Rat
Clone: BVD10-11C10
Isotype: IgG2a
Species Reactivity: Human
Immunogen: Recombinant Human IL-7

ELISA, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-7
Applications ELISA
Classification Monoclonal
Clone BVD10-11C10
Concentration 0.5mg/mL
Conjugate Biotin
Formulation Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Host Species Rat
Immunogen Recombinant Human IL-7
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.