Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-8 Mouse anti-Human, Brilliant Violet 510, Clone: G265-8, BD

Catalog No. BDB563311 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563311 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563311 Supplier BD Biosciences Supplier No. 563311
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The G265-8 monoclonal antibody specifically binds to both the 72 and 77 amino acid isoforms of human Interleukin-8 (IL-8). IL-8 is secreted as an 8-9 kDa, non-glycosylated proinflammatory chemokine protein also known as chemokine (C-X-C motif) ligand 8 (CXCL8). IL-8 is synthesized as a 99 amino acid precursor that is proteolytically processed into several isoforms. The 72 amino acid isoform is produced by monocytes, macrophages, granulocytes, epithelial cells, and fibroblasts in response to pro-inflammatory stimuli including cytokines and microbial agents. It is also expressed by endothelial cells, fibroblasts, keratinocytes, lymphocytes, and a variety of tumor cells. In response to IL-4, IL-10 and TGFβ, the cellular production of IL-8 is inhibited. IL-8 is crucial for the activation and recruitment of neutrophils to inflammatory sites. IL-8 is also a chemoattractant for basophils and T-lymphocytes. IL-8 possesses angiogenic activity and can be associated with tumor angiogenesis and metastasis. The 77 amino acid IL-8 isoform is primarily produced by endothelial cells. This larger isoform is reportedly a less potent neutrophil activator than the 72 amino acid isoform. IL-8 binds to and signals through two G-protein-coupled receptors, IL-8RA (CXCR1/CD181) and IL-8RB (CXCR2/CD182).

The antibody was conjugated to BD Horizon™ BV510 which is part of the BD Horizon™ Brilliant Violet™ family of dyes. With an Ex Max of 405-nm and Em Max at 510-nm, BD Horizon™ BV510 can be excited by the violet laser and detected in the BD Horizon™ V500 (525/50-nm) filter set. BD Horizon™ BV510 conjugates are useful for the detection of dim markers off the violet laser.

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-8
Applications Flow Cytometry
Classification Monoclonal
Clone G265-8
Conjugate Brilliant Violet 510
Description IL8; CXCL8; GCP-1; LYNAP; MDNCF; MONAP; NAP-1; emoctakin
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Recombinant Human IL-8
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.