Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-8 Mouse anti-Human, NA/LE, Unlabeled, Clone: 2A2, BD

Catalog No. BDB554726 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554726 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554726 Supplier BD Biosciences Supplier No. 554726
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 2A2 antibody reacts with both the 72 and 77 amino acid forms of human IL-8. The immunogen used to produce the 2A2 hybridoma was E. coli-expressed recombinant human IL-8 (77 amino acid form). This antibody shows no cross-reactivity with any other cytokines or chemokines tested (e.g., human IL-1α, IL-1β, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-12 (p40 or p70), IL-13, IL-15, eotaxin, GRO, IFN- γ, MCP-1, MCP-2, MCP-3, MIP-1α, MIP-1β, NAP-2, PF4, RANTES, SCF, TNF, LT-α)

Host Species: Mouse
Clone: 2A2
Isotype: IgG1
Species Reactivity: Human
Immunogen: Recombinant human IL-8

Functional Assay, Neutralization, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-8
Applications ELISA
Classification Monoclonal
Clone 2A2
Concentration 1.0mg/mL
Conjugate Unconjugated
Formulation No azide/low endotoxin: Aqueous buffered solution containing no preservative, 0.2μm sterile filtered. Endotoxin level is ≤0.01 EU/μg (≤0.001 ng/μg) of protein as determined by the LAL assay.
Host Species Mouse
Immunogen Recombinant human IL-8
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.