Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-8 Mouse anti-Human, Unlabeled, Clone: G265-5, BD

Catalog No. BDB554716 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554716 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554716 Supplier BD Biosciences Supplier No. 554716
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The G265-8 antibody reacts with both the 72 and 77 amino acid forms of human interleukin-8 (IL-8). The immunogen used to produce the G265-8 hybridoma was recombinant human IL-8.
This antibody is routinely tested by ELISA as the capture. Other applications were tested at BD Biosciences Pharmingen during antibody development or reported in the literature.

Host Species: Rat
Clone: G265-5
Isotype: IgG2b κ
Species Reactivity: Human
Immunogen: Recombinant Human IL-8

ELISA, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-8
Applications ELISA
Classification Monoclonal
Clone G265-5
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Recombinant Human IL-8
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG2b κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.