Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-9 Hamster anti-Mouse, NA/LE, Unlabeled, Clone: D9302C12, BD

Catalog No. BDB554472 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554472 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554472 Supplier BD Biosciences Supplier No. 554472

Hamster Monoclonal Antibody

The D9302C12 antibody reacts with mouse interleukin-9 (IL-9). The immunogen used to generate the D9302C12 hybridoma was recombinant mouse IL-9. The NA/LE format of the D9302C12 antibody (Mfr. No. 554472) is useful for neutralization of mouse IL-9 bioactivity. This antibody preparation is tested in bioassay to verify neutralization activity and LAL assay to assess endotoxin level.

Host Species: Hamster
Clone: D9302C12
Isotype: IgG2 κ
Species Reactivity: Mouse
Immunogen: Recombinant Mouse IL-9

Functional Assay, Neutralization

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-9
Applications ELISA
Classification Monoclonal
Clone D9302C12
Concentration 1.0mg/mL
Conjugate Unconjugated
Description IL-9; Interleukin-9; MEA; P40; T-cell growth factor P40; TCGF III
Formulation No azide/low endotoxin: Aqueous buffered solution containing no preservative, 0.2μm sterile filtered. Endotoxin level is ≤0.01 EU/μg (≤0.001 ng/μg) of protein as determined by the LAL assay.
Host Species Hamster
Immunogen Recombinant Mouse IL-9
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG2 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.