Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-9 Mouse anti-Human, Biotin, Clone: MH9A3, BD

Catalog No. BDB558089 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558089 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558089 Supplier BD Biosciences Supplier No. 558089

Mouse Monoclonal Antibody

Host Species: Mouse
Clone: MH9A3
Isotype: IgG1, κ
Species Reactivity: Hu
Immunogen: Human IL-9 Recombinant Protein

ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-9
Applications ELISA
Classification Monoclonal
Clone MH9A3
Concentration 0.5mg/mL
Conjugate Biotin
Host Species Mouse
Immunogen Human IL-9 Recombinant Protein
Quantity 0.5 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.