Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-9 Mouse anti-Human, PE, Clone: MH9A3, BD

Catalog No. BDB560807 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560807 100 Tests
BDB560814 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB560807 Supplier BD Biosciences Supplier No. 560807

Mouse Monoclonal Antibody

The MH9A3 monoclonal antibody specifically binds to human interleukin-9 (IL-9). Human IL-9 is a multifunctional cytokine and a member of the type I cytokine (hematopoietin) family that includes IL-2, IL-4, IL-7, IL-15 and IL-21. This cytokine is encoded by the IL9 gene that is resident on chromosome 5q31.1. IL-9 is expressed by activated CD4-positive T helper cells, by some transformed T cells and by eosinophils, mast cells and neutrophils. IL-9 induces the proliferation, differentiation, and effector function of various cell types including T lymphocytes, B lymphocytes, mast cells, eosinophils, neutrophils, hematopoietic cells and epithelial cells. It potentiates the interleukin-4-induced IgM, IgG and IgE responses by human B lymphocytes. IL-9 has been implicated in human allergic disorders such as asthma and malignancies such as Hodgkin’s disease. IL-9 exerts its biological activities through binding to the surface IL-9 receptor (IL-9R) complex comprised of the IL-9R alpha subunit (IL-9R α ; CD129) and the common cytokine receptor gamma subunit ( γc; CD132). IL-9 signaling through its receptor includes activation of the Janus kinases 1 and 3 ( JAK1 and JAK3) and activation of Signal transducer and activator of transcription 1, 3 and 5 factors (STAT1, STAT3 and STAT5).

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-9
Applications Flow Cytometry
Classification Monoclonal
Clone MH9A3
Conjugate PE
Description IL9; IL-9; interleukin-9; HP40; P40
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human IL-9 Recombinant Protein
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.