Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-9 Armenian Hamster anti-Mouse, PerCP-Cy5.5, Clone: D9302C12, BD

Catalog No. BDB561492 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561492 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB561492 Supplier BD Biosciences Supplier No. 561492

Armenian Hamster Monoclonal Antibody

The D9302C12 monoclonal antibody specifically binds to the multifunctional mouse cytokine, Interleukin-9 (IL-9). IL-9 is a 126 amino acid-long glycoprotein that is produced by various subsets of activated CD4+ T cells. IL-9 acts on target cells by binding to and signaling through the heterodimeric IL-9 receptor (IL-9R) complex that is comprised of transmembrane IL-9 receptor alpha (IL-9R α) and common gamma chain (γc) subunits. IL-9 can promote the survival, growth, proliferation and/or differentiation of various cell types including thymocytes, T cells, B cells, mast cells, and hematopoietic progenitor cells. IL-9 can augment IL-4-induced IgE and IgG1 production from lipopolysaccharide-primed mouse B cells and induce granzyme and high-affinity IgE receptor gene expression by mouse T helper cell clones and mast cell lines. IL-9 plays an important role in vivo in helminth elimination. The D9302C12 antibody neutralizes mouse IL-9 bioactivity.

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-9
Applications Flow Cytometry
Classification Monoclonal
Clone D9302C12
Concentration 0.2mg/mL
Conjugate PerCP-Cyanine5.5
Description IL-9; Interleukin-9; MEA; P40; T-cell growth factor P40; TCGF III
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Armenian Hamster
Immunogen Mouse IL-9 Recombinant Protein
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.