Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

interleukin 13, Mouse, Clone: 7E4, Abnova™

Catalog No. 89000752 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-000-752 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-000-752 Supplier Abnova Corporation Supplier No. H00003596M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant IL13.

This gene encodes an immunoregulatory cytokine produced primarily by activated Th2 cells. This cytokine is involved in several stages of B-cell maturation and differentiation. It up-regulates CD23 and MHC class II expression, and promotes IgE isotype switching of B cells. This cytokine down-regulates macrophage activity, thereby inhibits the production of pro-inflammatory cytokines and chemokines. This cytokine is found to be critical to the pathogenesis of allergen-induced asthma but operates through mechanisms independent of IgE and eosinophils. This gene, IL3, IL5, IL4, and CSF2 form a cytokine gene cluster on chromosome 5q, with this gene particularly close to IL4. [provided by RefSeq

Specifications

Antigen interleukin 13
Applications ELISA, Western Blot
Classification Monoclonal
Clone 7E4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant IL13.
Formulation PBS with no preservative; pH 7.4
Gene IL13
Gene Accession No. P35225
Gene Alias ALRH/BHR1/IL-13/MGC116786/MGC116788/MGC116789/P600
Gene Symbols IL13
Host Species Mouse
Immunogen IL13 recombinant protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 3596
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.