Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-IL13RA2 Clone: 20000000000,

Catalog No. AP1145100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
AP1145100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP1145100UG Supplier MilliporeSigma™ Supplier No. AP1145100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects IL13RA2 Clone: 20000000000 in Human samples, and it is validated for ELISA, Immuno Blotting

IL13RA2 is closely related to IL13RA1, a subunit of the interleukin 13 receptor complex. This protein binds IL13 with high affinity, but lacks a cytoplasmic domain, and does not appear to function as a signal mediator. It is reported to play a role in the internalization of IL13.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL13RA2
Applications ELISA, Immunoblot, Immunoblot
Classification Monoclonal
Clone 20000000000
Conjugate Unconjugated
Formulation In PBS, pH 7.2.
Host Species Mouse
Immunogen A recombinant polypeptide corresponding to amino acids 27-127 of human IL13RA2, expressed as a GST fusion protein
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽50kDa IL13RA2 protein in HeLa cells.
Target Species Human
Content And Storage −20°C
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.