Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

interleukin 16 (lymphocyte chemoattractant factor), Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89017778 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-778 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-778 Supplier Abnova Corporation Supplier No. H00003603D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human IL16 protein.

The protein encoded by this gene is a pleiotropic cytokine that functions as a chemoattractant, a modulator of T cell activation, and an inhibitor of HIV replication. The signaling process of this cytokine is mediated by CD4. The product of this gene undergoes proteolytic processing, which is found to yield two functional proteins. The cytokine function is exclusively attributed to the secreted C-terminal peptide, while the N-terminal product may play a role in cell cycle control. Caspase 3 is reported to be involved in the proteolytic processing of this protein. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: MESHSRAGKSRKSAKFRSISRSLMLCNAKTSDDGSSPDEKYPDPFEISLAQGKEGIFHSSVQLADTSEAGPSSVPDLALASEAAQLQAAGNDRGKTCRRIFFMKESSTASSREKPGKLEAQSSNFLFPKACHQRARSNSTSVNPYCTREIDFPMTKKSAAPTDRQPYSLCSNRKSLSQQLDCPAGKAAGTSRPTRSLSTAQLVQPSGGLQASVISNIVLMKGQAKGLGFSIVGGKDSIYGPIGIYVKTIFAGGAAAADGRLQEGDEILELNGESMAGLTHQDALQKFKQAKKGLLTLTVRTRLTAPPSLCSHLSPPLCRSLSSSTCITKDSSSFALESPSAPISTAKPNYRIMVEVSLQKEAGVGLGIGLCSVPYFQCISGIFVHTLSPGSVAHLDGRLRCGDEIVEISDSPVHCLTLNEVYTILSHCDPGPVSIIVSRHPDPQKEEMEAQEVK

Specifications

Antigen interleukin 16 (lymphocyte chemoattractant factor)
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human IL16 protein.
Formulation PBS with no preservative; pH 7.4
Gene IL16
Gene Accession No. BC040272
Gene Alias FLJ16806/FLJ42735/FLJ44234/HsT19289/IL-16/LCF/prIL-16
Gene Symbols IL16
Host Species Rabbit
Immunogen IL16 (AAH40272.1, 1 a.a. ∼ 454 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cytokines
Primary or Secondary Primary
Gene ID (Entrez) 3603
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.