Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL1R2 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA524525
Description
The protein encoded by this gene is a cytokine receptor that belongs to the interleukin 1 receptor family. This protein binds interleukin alpha (IL1A), interleukin beta (IL1B), and interleukin 1 receptor, type I(IL1R1/IL1RA), and acts as a decoy receptor that inhibits the activity of its ligands. Interleukin 4 (IL4) is reported to antagonize the activity of interleukin 1 by inducing the expression and release of this cytokine. This gene and three other genes form a cytokine receptor gene cluster on chromosome 2q12. Two alternatively spliced transcript variants encoding the same protein have been reported.
Specifications
IL1R2 | |
Polyclonal | |
IL1R2 | |
CD121b, IL1RB, CD121 antigen-like family member B, CDw121b, IL-1 type II receptor, IL-1R-2, IL-1R-beta, IL-1RT-2, IL-1RT2, antigen CDw121b, interleukin-1 receptor beta, interleukin-1 receptor type 2, interleukin-1 receptor type II, type II interleukin-1 receptor, beta, ILR2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7850 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
Unconjugated | |
P27930 | |
IL1R2 | |
KLH conjugated synthetic peptide between 273-300 amino acids from the central region of human IL1R2 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction