Learn More
Description
Specifications
Specifications
| Antigen | IL1RL1 |
| Applications | Functional Assay, Flow Cytometry |
| Classification | Monoclonal |
| Clone | RMST2-2 |
| Conjugate | Unconjugated |
| Formulation | Purified rat monoclonal IgG2aκ in buffer containing PBS without preservatives. |
| Gene Accession No. | NP_001020773 |
| Gene Symbols | Il1rl1, Ly84, St2, Ste2 |
| Host Species | Rat |
| Immunogen | Mouse ST-2/IL1RL1. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.