Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

interleukin 27, Mouse, Clone: 3F12, Abnova™

Catalog No. 89014352 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-014-352 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-014-352 Supplier Abnova Corporation Supplier No. H00246778M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant IL27.

The protein encoded by this gene is one of the subunits of a heterodimeric cytokine complex. This protein is related to interleukin 12A (IL12A). It interacts with Epstein-Barr virus induced gene 3 (EBI3), a protein similar to interleukin 12B (IL12B), and forms a complex that has been shown to drive rapid expansion of naive but not memory CD4(+) T cells. The complex is also found to synergize strongly with interleukin 12 to trigger interferon gamma (IFNG) production of naive CD4(+) T cells. The biological effects of this cytokine are mediated by the class I cytokine receptor (WSX1/TCRR). [provided by RefSeq]

Sequence: RKGLLPGALGSALQGPAQVSWPQLLSTYRLLHSLELVLSRAVRELLLLSKAGHSVWPLGFPTLSPQP

Specifications

Antigen interleukin 27
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 3F12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant IL27.
Formulation PBS with no preservative; pH 7.4
Gene IL27
Gene Accession No. NM_145659
Gene Alias IL-27/IL-27A/IL27p28/IL30/MGC71873/p28
Gene Symbols IL27
Host Species Mouse
Immunogen IL27 (NP_663634, 177 a.a. ∼ 243 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle
Primary or Secondary Primary
Gene ID (Entrez) 246778
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.