Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

interleukin 31 receptor A (A01), Mouse anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89004787 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-787 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-787 Supplier Abnova Corporation Supplier No. H00133396A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant IL31RA.

IL31RA is related to gp130 (IL6ST; MIM 600694), the common receptor subunit for IL6 (MIM 147620)-type cytokines. Oncostatin M receptor (OSMR; MIM 601743) and IL31RA form the heterodimeric receptor through which IL31 (MIM 609509) signals. Expression of IL31RA and OSMR mRNA is induced in activated monocytes, and both mRNAs are constitutively expressed in epithelial cells (Dillon et al., 2004 [PubMed 15184896]).[supplied by OMIM

Sequence: LPAKPENISCVYYYRKNLTCTWSPGKETSYTQYTVKRTYAFGEKHDNCTTNSSTSENRASCSFFLPRITIPDNYTIEVEAENGDGVIKSHMTYWRLENIA

Specifications

Antigen interleukin 31 receptor A
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant IL31RA.
Formulation 50% glycerol
Gene IL31RA
Gene Accession No. NM_139017
Gene Alias CRL/CRL3/GLM-R/GLMR/GPL/IL-31RA/MGC125346/PRO21384
Gene Symbols IL31RA
Host Species Mouse
Immunogen IL31RA (NP_620586, 21 a.a. ∼ 120 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Apoptosis
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 133396
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.