Learn More
IL6, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human IL6 peptide using ARM Technology.
Supplier: Abnova Corporation H00003569K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
This gene encodes a cytokine that functions in inflammation and the maturation of B cells. The protein is primarily produced at sites of acute and chronic inflammation, where it is secreted into the serum and induces a transcriptional inflammatory response through interleukin 6 receptor, alpha. The functioning of this gene is implicated in a wide variety of inflammation-associated disease states, including suspectibility to diabetes mellitus and systemic juvenile rheumatoid arthritis. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
IL6 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
IL6 | |
IL6 | |
A synthetic peptide of human IL6 | |
RUO | |
3569 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
interleukin 6 (interferon, beta 2) | |
IL6 | |
BSF2, HGF, HSF, IFNB2, IL-6 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.