Learn More
interleukin 7, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human IL7 protein.
Supplier: Abnova Corporation H00003574D01P
Description
The protein encoded by this gene is a cytokine important for B and T cell development. This cytokine and the hepatocyte growth factor (HGF) form a heterodimer that functions as a pre-pro-B cell growth-stimulating factor. This cytokine is found to be a cofactor for V(D)J rearrangement of the T cell receptor beta (TCRB) during early T cell development. This cytokine can be produced locally by intestinal epithelial and epithelial goblet cells, and may serve as a regulatory factor for intestinal mucosal lymphocytes. Knockout studies in mice suggested that this cytokine plays an essential role in lymphoid cell survival. [provided by RefSeq
Sequence: MFHVSFRYIFGLPPLILVLLPVASSDCDIEGKDGKQYESVLMVSIDQLLDSMKEIGSNCLNNEFNFFKRHICDANKEGMFLFRAARKLRQFLKMNSTGDFDLHLLKVSEGTTILLNCTGQVKGRKPAALGEAQPTKSLEENKSLKEQKKLNDLCFLKRLLQEIKTCWNKILMGTKEHSpecifications
interleukin 7 | |
Polyclonal | |
Rabbit polyclonal antibody raised against a full-length human IL7 protein. | |
IL7 | |
IL-7 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
Antibody |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_000880 | |
IL7 | |
IL7 (AAH47698.1, 1 a.a. ∼ 177 a.a) full-length human protein. | |
100 μg | |
Cell Cycle | |
3574 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.