Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-ILT4/CD85d, Fluorescein, Clone: 287219, R&D Systems™
Specifications
Antigen | ILT4/CD85d/LILRB2 |
---|---|
Clone | 287219 |
Dilution | Flow Cytometry 10 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB2078F025
|
R&D Systems
FAB2078F025 |
25 Tests | N/A | N/A | N/A | ||||
FAB2078F100
|
R&D Systems
FAB2078F100 |
100 Tests | N/A | N/A | N/A | ||||
Description
Applications:
Flow Cytometry
Specifications
ILT4/CD85d/LILRB2 | |
Flow Cytometry 10 uL/10^6 cells | |
Monoclonal | |
Purified | |
RUO | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
CD85 antigen-like family member D, CD85d, CD85d antigen, Ig-like transcript 4, ILT4, ILT-4, ILT4CD85d, Immunoglobulin-like transcript 4, Leukocyte immunoglobulin-like receptor 2, leukocyte immunoglobulin-like receptor subfamily B member 2, leukocyte immunoglobulin-like receptor subfamily B member 2 soluble isoform, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR2, LIR2CD85D, LIR-2subfamily A (with TM domain), member 6, member 2, MIR10, MIR10LILRA6, Monocyte/macrophage immunoglobulin-like receptor 10 | |
LILRB2 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Detects human ILT4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) ILT1, rhILT2, rhILT3, rhILT6, rhILT7, rhILT11, rhLIR6 or rhLIR8 is observed. |
287219 | |
Flow Cytometry | |
Fluorescein | |
Mouse | |
Human | |
ACT64556 | |
10288 | |
Mouse myeloma cell line NS0-derived recombinant human ILT4, Gly24-His458, Accession # ACT64556 | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title