Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                Human LILRB2/CD85d/ILT4 PE-conjugated Antibody, R&D Systems™
                                
                                
                                
                                
                            
                            
                            
                                
                                    
Mouse Monoclonal Antibody has been used in 4 publications
$164.79 - $460.95
Specifications
| Antigen | ILT4/CD85d/LILRB2 | 
|---|---|
| Clone | 287219 | 
| Dilution | Flow Cytometry 10 uL/10^6 cells | 
| Applications | Flow Cytometry | 
| Classification | Monoclonal | 
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
                
                
                    
                        FAB2078P025
                    
                    
                        
                    
                    
                        ![]()  | 
            
                
                    
                        R&D Systems
                         FAB2078P025  | 
                
            
            
            
            
            
                
                    25 Tests | 
                    
                        
                        
                            
                             
                                        
                                            
                                                
                                                
                                                
                                            
                                        
                                        
                                            
                                            
                                                
                                                
                                            
                                        
                                        
                                            
                                                
                                                Each for $164.79
                                                 
                                
                             | 
                
                    
                        
                             | 
                    
                
                
                    |||||
                
                
                    
                        FAB2078P100
                    
                    
                        
                    
                    
                        ![]()  | 
            
                
                    
                        R&D Systems
                         FAB2078P100  | 
                
            
            
            
            
            
                
                    100 Tests | 
                    
                        
                        
                            
                             
                                        
                                            
                                                
                                                
                                                
                                            
                                        
                                        
                                            
                                            
                                                
                                                
                                            
                                        
                                        
                                            
                                                
                                                Each for $460.95
                                                 
                                
                             | 
                
                    
                        
                             | 
                    
                
                
                    |||||
Description
ILT4/CD85d/LILRB2 Monoclonal specifically detects ILT4/CD85d/LILRB2 in Human samples. It is validated for Flow Cytometry.Specifications
| ILT4/CD85d/LILRB2 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CD85 antigen-like family member D, CD85d, CD85d antigen, Ig-like transcript 4, ILT4, ILT-4, ILT4CD85d, Immunoglobulin-like transcript 4, Leukocyte immunoglobulin-like receptor 2, leukocyte immunoglobulin-like receptor subfamily B member 2, leukocyte immunoglobulin-like receptor subfamily B member 2 soluble isoform, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR2, LIR2CD85D, LIR-2subfamily A (with TM domain), member 6, member 2, MIR10, MIR10LILRA6, Monocyte/macrophage immunoglobulin-like receptor 10 | |
| LILRB2 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects human ILT4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) ILT1, rhILT2, rhILT3, rhILT6, rhILT7, rhILT11, rhLIR6 or rhLIR8 is observed. | 
| 287219 | |
| Flow Cytometry | |
| R-PE | |
| Mouse | |
| Human | |
| ACT64556 | |
| 10288 | |
| Mouse myeloma cell line NS0-derived recombinant human ILT4 Gly24-His458 Accession # ACT64556 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | 
Spot an opportunity for improvement?Share a Content Correction
            
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title