Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human LILRB3/CD85a/ILT5 APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
$151.20 - $456.00
Specifications
Antigen | ILT5/CD85a/LILRB3 |
---|---|
Clone | 222821 |
Dilution | Flow Cytometry 10 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB1806A025
![]() |
R&D Systems
FAB1806A025 |
25 Tests |
Each for $151.20
|
|
|||||
FAB1806A100
![]() |
R&D Systems
FAB1806A100 |
100 Tests |
Each for $456.00
|
|
|||||
Description
ILT5/CD85a/LILRB3 Monoclonal specifically detects ILT5/CD85a/LILRB3 in Human samples. It is validated for Flow Cytometry.Specifications
ILT5/CD85a/LILRB3 | |
Flow Cytometry 10 uL/10^6 cells | |
Monoclonal | |
Purified | |
RUO | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
CD85 antigen-like family member A, CD85a, CD85a antigen, HL9Immunoglobulin-like transcript 5, ILT5, ILT-5, ILT5CD85a, Leukocyte immunoglobulin-like receptor 3, leukocyte immunoglobulin-like receptor subfamily B member 3, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR3, LIR3CD85A, LIR-3MGC138403, member 3, Monocyte inhibitory receptor HL9, PIRB | |
LILRB3 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Detects human ILT5/CD85a in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) ILT1, rhILT2, rhILT3, rhILT4, rhILT6, rhILT7 or rhILT11 is observed. |
222821 | |
Flow Cytometry | |
APC | |
Mouse | |
Human | |
AAB68668 | |
11025 | |
Mouse myeloma cell line NS0-derived recombinant human ILT5/CD85a Gly24-Glu443 Accession # AAB68668 | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title