Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Indolamine (Indoleamine) 2, Polyclonal,

Catalog No. AB9252MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB9252MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB9252MI Supplier MilliporeSigma™ Supplier No. AB9252

Specifications

Antigen Indolamine (Indoleamine) 2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Sheep serum. Liquid containing 0.02% sodium azide.
Gene Symbols INDO; IDO; EC 1.13.11.52
Host Species Sheep
Immunogen Recombinant human IDO.
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_002164.4
Target Species Human
Content And Storage −20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.