Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza A Mouse anti-Human, Biotin, Clone: A1, MilliporeSigma™

Catalog No. MAB8257B5MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8257B5MI 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB8257B5MI Supplier MilliporeSigma Supplier No. MAB8257B5
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Influenza A Antibody, nucleoprotein, clone A1 is an antibody against Influenza A for use in immunofluoresence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Influenza A
Applications Immunofluorescence
Classification Monoclonal
Clone A1
Conjugate Biotin
Formulation Biotin conjugated purified immunoglobulin. Liquid in 0.01M PBS, pH=7.1, 0.1% Sodium Azide with 15mg/mL BSA as stabilizer.
Host Species Mouse
Immunogen Influenza A
Quantity 500 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 12 months. Protect from Light.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.