Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza A Mouse anti-Human, Clone: A3, MilliporeSigma™

Catalog No. MAB82585MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB82585MI 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB82585MI Supplier MilliporeSigma Supplier No. MAB82585
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Influenza A Antibody, nucleoprotein, clone A3 detects level of Influenza A and has been published and validated for use in immunofluoresence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Influenza A
Applications Immunofluorescence
Classification Monoclonal
Clone A3
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid in 0.02M phosphate buffer, 0.25M sodium chloride, pH 7.6 with 0.1% sodium azide
Host Species Mouse
Immunogen Influenza A
Quantity 500 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.