Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza A Mouse anti-Human, FITC, Clone: A1, MilliporeSigma™

Catalog No. MAB8257FMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8257FMI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB8257FMI Supplier MilliporeSigma Supplier No. MAB8257F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Influenza A Antibody, nucleoprotein, clone A1, FITC-conjugated is validated for use in IF for the detection of Influenza A.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Influenza A
Applications Immunofluorescence
Classification Monoclonal
Clone A1
Conjugate FITC
Formulation Purified, FITC conjugated. Liquid in 0.02M phosphate buffer, 0.25M sodium chloride, pH 7.6 with 15mg/mL BSA and 0.1% sodium azide
Host Species Mouse
Immunogen Influenza A
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.