Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza A Goat anti-Human, Polyclonal, MilliporeSigma™

Catalog No. AB1074MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB1074MI 1 Unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB1074MI Supplier MilliporeSigma Supplier No. AB1074
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Anti-Influenza A Antibody is an antibody against Influenza A for use in ELISA, immunofluoresence, immunohistochemistry and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Influenza A
Applications ELISA, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Protein A Purified goat immunoglobulin in PBS (0.01 M, pH 7.2) with 0.1% sodium azide as a preservative.
Host Species Goat
Immunogen Influenza A-USSR (H1N1).
Purification Method Protein A purfied
Quantity 1 Unit
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for 3 months or at −20°C in aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.