Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza A H3N2, Polyclonal, Invitrogen™

Catalog No. PIPA127282
Change view
Click to view available options
Quantity:
1 mL
Catalog No. Quantity
PIPA127282 1 mL
1 options

Catalog No. PIPA127282

Supplier: Thermo Scientific PA127282

Goat Polyclonal Antibody

Influenza viruses consists of three types (Influenza A, B, and C), which make up three of the five genera in the Orthomyxoviridae family. Commonly know as the flu, influenza strains can affect both birds and mammals. Influenza strains form roughly spherical particles and are similar in composition with viral envelope containing two main types of glycoproteins (hemagglutinin and neuraminidase), wrapped around the central core of the viral RNA genome and other viral proteins that package and protect the RNA. Unusually for a virus, the influenzavirus genome is not a single piece of nucleic acid; rather a seven to eight segmented fragments of negative-sense RNA with each fragment expressing different viral proteins. Influenza spreads seasonally, resulting in about three to five million yearly cases of severe illness and about 250,000 to 500,000 deaths annually or into the millions in pandemic years.

PA1-27282 detects Influenza A (H3N2) from virus samples.PA1-27282 has been successfully used in immunofluorescence procedures.The PA1-27282 immunogen is Influenza A strain Texas 1/77 (H3N2).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Influenza A H3N2
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 4mg/mL
Conjugate Unconjugated
Gene Alias Influenza H3N2, H3N2
Host Species Goat
Immunogen Influenza A strain Texas 1/77 (H3N2).
Purification Method IgG fraction
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.