Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza B Mouse anti-Human, Biotin, Clone: 22D5-12-13, MilliporeSigma™

Catalog No. MAB8671B5MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8671B5MI 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB8671B5MI Supplier MilliporeSigma Supplier No. MAB8671B5
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Influenza B Antibody, Core, clone 22D5-12-13 detects level of Influenza B and has been published and validated for use in immunofluoresence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Influenza B
Applications Immunofluorescence
Classification Monoclonal
Clone 22D5-12-13
Conjugate Biotin
Formulation Biotin conjugated purified immunoglobulin. Liquid in 0.01M PBS, pH=7.1, 0.1% Sodium Azide with 15mg/mL BSA as stabilizer.
Host Species Mouse
Quantity 500 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year. Protect from light.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.