Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Influenza B Mouse anti-Human, FITC, Clone: B2, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB8259F
Description
Anti-Influenza B Antibody, clone B2, FITC-conjugated is an antibody against Influenza B for use in immunofluoresence.
Specifications
Influenza B | |
Monoclonal | |
FITC | |
Mouse | |
100 μg | |
Human | |
Purified |
Immunofluorescence | |
B2 | |
Purified, FITC conjugated. Liquid in 0.02M phosphate buffer, 0.25M sodium chloride, pH 7.6 with 15mg/mL BSA and 0.1% sodium azide | |
Influenza B | |
Primary | |
2-8°C. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction