Learn More
inhibitor of growth family, member 2 (B01), Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human ING2 protein.
Supplier: Abnova Corporation H00003622B01
Description
This gene is a member of the inhibitor of growth (ING) family. Members of the ING family associate with and modulate the activity of histone acetyltransferase (HAT) and histone deacetylase (HDAC) complexes and function in DNA repair and apoptosis. [provided by RefSeq
Sequence: MLGQQQQQLYSSAALLTGERSRLLTCYVQDYLECVESLPHDMQRNVSVLRELDNKYQETLKEIDDVYEKYKKEDDLNQKKRLQQLLQRALINSQELGDEKIQIVTQMLELVENRARQMELHSQCFQDPAESERASDKAKMDSSQPERSSRRPRRQRTSESRDLCHMANGIEDCDDQPPKEKKSKSAKKKKRSKAKQEREASPVEFAIDPNEPTYCLCNQVSYGEMIGCDNEQCPIEWFHFSCVSLTYKPKGKWYCPKCRGDNEKTMGKSTEKTKKDRRSRSpecifications
inhibitor of growth family, member 2 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human ING2 protein. | |
ING2 | |
ING1L/p33ING2 | |
Mouse | |
50 μL | |
Yes | |
3622 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Immunofluorescence, Western Blot | |
Unconjugated | |
No additive | |
BC030128 | |
ING2 | |
ING2 (AAH30128, 1 a.a. ∼ 280 a.a) full-length human protein. | |
RUO | |
Primary | |
Human | |
Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.