Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-INHBA, Polyclonal

Catalog No. CB1030100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
CB1030100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CB1030100UG Supplier MilliporeSigma™ Supplier No. CB1030100UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects INHBA Clone: in Human, Mouse samples, and it is validated for Immuno Blotting

The inhibin beta A subunit joins the alpha subunit to form a pituitary FSH secretion inhibitor. Inhibin has been shown to regulate gonadal stromal cell proliferation negatively and to have tumor-suppressor activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen INHBA
Applications Immunoblot
Classification Polyclonal
Conjugate Unconjugated
Formulation In PBS, pH 7.2.
Host Species Rabbit
Immunogen Full-length human INHBA (aa 1-426)
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽50kDa INHBA protein in mouse liver tissue.
Target Species Human, Mouse
Content And Storage −20°C
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.