Learn More
iNOS/NOS II Rabbit anti-Mouse, Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABN26
Description
Detect iNOS/NOS II using this Anti-iNOS/NOS II Antibody, NT validated for use in western blot, immunoprecipitation, immunohistochemistry (paraffin).
Specifications
iNOS/NOS II | |
Polyclonal | |
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
Rabbit | |
Affinity Purified | |
Primary | |
Human, Mouse |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Nos2; Inosl | |
GST-tagged recombinant protein corresponding to the N-terminus of mouse iNOS/NOS II. | |
100 μg | |
NP_035057 | |
2-8°C for 12 months |